Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000926.4 | NP_000917.3 | 2 | 2517 | ||
NM_001202474.3 | NP_001189403.1 | 2 | 1767 | |||
NM_001271161.2 | NP_001258090.1 | 2 | 1767 | |||
NM_001271162.1 | NP_001258091.1 | 2 | 437 | |||
NR_073141.2 | 2 | 1767 | ||||
NR_073142.2 | 2 | 1767 | ||||
NR_073143.2 | 2 | 1767 | ||||
XM_006718858.3 | XP_006718921.1 | 2 | 3367 | |||
XM_011542869.2 | XP_011541171.1 | 2 | 3385 | |||
GenBank mRNA | AB084248.1 | - | 2 | 1774 | ||
AB085683.1 | - | 2 | 1774 | |||
AB085843.1 | - | 2 | 1774 | |||
AB085844.1 | - | 2 | 1774 | |||
AB085845.1 | - | 2 | 1774 | |||
AF016381.1 | - | 2 | 1949 | |||
AK304853.1 | - | 2 | 437 | |||
AY382151.1 | - | 1 | 133 | |||
M15716.1 | - | 2 | 1949 | |||
X51730.1 | - | 2 | 3228 |