Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000927.4 | NP_000918.2 | 14 | 1882 | ||
GenBank mRNA | AB208970.1 | - | 12 | 1352 | ||
AF016535.1 | - | 13 | 1505 | |||
AK290159.1 | - | 14 | 1883 | |||
BC130424.1 | - | 13 | 1714 | 40146455 | ||
BQ882401.1 | - | 6 | 655 | |||
EU852583.1 | - | 12 | 1389 | |||
EU854148.1 | - | 11 | 1197 | |||
M14758.1 | - | 14 | 1813 |