Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000927.4 | NP_000918.2 | 21 | 2954 | ||
GenBank mRNA | AB208970.1 | - | 19 | 2424 | ||
AF016535.1 | - | 20 | 2577 | |||
AK290159.1 | - | 21 | 2955 | |||
BC130424.1 | - | 20 | 2786 | 40146455 | ||
BQ720763.1 | - | 2 | 129 | |||
EU852583.1 | - | 19 | 2461 | |||
EU854148.1 | - | 18 | 2269 | |||
M14758.1 | - | 21 | 2885 |