Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000293.2 | NP_000284.1 | 9 | 887 | ||
NM_001031835.2 | NP_001027005.1 | 10 | 999 | |||
XM_005255983.4 | XP_005256040.1 | 9 | 850 | |||
XM_005255984.4 | XP_005256041.1 | 10 | 1150 | |||
XM_017023282.1 | XP_016878771.1 | 3 | 144 | |||
XM_017023283.1 | XP_016878772.1 | 10 | 972 | |||
XM_017023284.1 | XP_016878773.1 | 9 | 853 | |||
GenBank mRNA | AK309226.1 | - | 9 | 850 | ||
BC033657.1 | - | 10 | 978 | 3913693 | ||
CR936710.1 | - | 10 | 1033 | |||
X84908.1 | - | 9 | 859 |