Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000295.4 | NP_000286.3 | 2 | 817 | ||
NM_001002235.2 | NP_001002235.1 | 2 | 796 | |||
NM_001002236.2 | NP_001002236.1 | 4 | 1110 | |||
NM_001127700.1 | NP_001121172.1 | 2 | 833 | |||
NM_001127701.1 | NP_001121173.1 | 4 | 1129 | |||
NM_001127702.1 | NP_001121174.1 | 3 | 937 | |||
NM_001127703.1 | NP_001121175.1 | 4 | 1092 | |||
NM_001127704.1 | NP_001121176.1 | 4 | 1089 | |||
NM_001127705.1 | NP_001121177.1 | 4 | 1107 | |||
NM_001127706.1 | NP_001121178.1 | 3 | 900 | |||
NM_001127707.1 | NP_001121179.1 | 3 | 897 | |||
XM_017021370.1 | XP_016876859.1 | 3 | 997 | |||
GenBank mRNA | AK026174.1 | - | 2 | 599 | ||
AK315637.1 | - | 3 | 853 | |||
BC011991.1 | - | 2 | 788 | 3859644 | ||
BC015642.2 | - | 2 | 583 | 4644658 | ||
BC070163.1 | - | 3 | 523 | |||
BT019455.1 | - | 1 | 556 | |||
BX161449.1 | - | 2 | 723 | |||
BX247968.1 | - | 4 | 1037 | |||
BX248002.1 | - | 1 | 565 | |||
BX248257.1 | - | 2 | 590 | |||
DQ682455.1 | - | 1 | 556 | |||
GU727620.1 | - | 1 | 557 | |||
K01396.1 | - | 1 | 575 | |||
KU178257.1 | - | 1 | 556 | |||
M11465.1 | - | 1 | 567 | |||
X01683.1 | - | 1 | 594 | |||
X17122.1 | - | 1 | 135 |