Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002646.3 | NP_002637.3 | 9, 10 | 2048 | ||
XM_005245257.2 | XP_005245314.1 | 8, 9 | 2113 | |||
XM_005245258.3 | XP_005245315.1 | 8, 9 | 2113 | |||
XM_011509630.2 | XP_011507932.1 | 10, 11 | 2907 | |||
XM_011509631.2 | XP_011507933.1 | 9, 10 | 2224 | |||
XM_011509633.2 | XP_011507935.1 | 8, 9 | 2113 | |||
XM_017001473.1 | XP_016856962.1 | 9, 10 | 3043 | |||
XM_017001474.1 | XP_016856963.1 | 9, 10 | 2246 | |||
XM_017001475.1 | XP_016856964.1 | 7, 8 | 994 | |||
GenBank mRNA | AB210006.1 | - | 7, 8 | 1602 | ||
BC132876.1 | - | 9, 10 | 2020 | 40146898 | ||
BC136645.1 | - | 9, 10 | 2020 | 9020635 | ||
BC144342.1 | - | 9, 10 | 2020 | 9052862 | ||
Y11312.1 | - | 9, 10 | 2084 | |||
Y13892.1 | - | 7, 8 | 1585 |