Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308020.1 | NP_001294949.1 | 13 | 1453 | ||
NM_002647.3 | NP_002638.2 | 14 | 1642 | |||
XM_011526031.2 | XP_011524333.1 | 15 | 1807 | |||
GenBank mRNA | AK090567.1 | - | 1 | 2176 | ||
AK298517.1 | - | 13 | 1397 | |||
BC033004.2 | - | 14 | 1592 | 4823337 | ||
BC053651.1 | - | 14 | 1572 | 5517878 | ||
Z46973.1 | - | 14 | 1595 |