Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308020.1 | NP_001294949.1 | 21, 22 | 2333 | ||
NM_002647.3 | NP_002638.2 | 22, 23 | 2522 | |||
XM_011526031.2 | XP_011524333.1 | 23, 24 | 2687 | |||
GenBank mRNA | AK090567.1 | - | 9, 10 | 3056 | ||
AK298517.1 | - | 21, 22 | 2277 | |||
BC033004.2 | - | 22, 23 | 2472 | 4823337 | ||
BC053651.1 | - | 22, 23 | 2452 | 5517878 | ||
CA945171.1 | - | 3, 4 | 594 |