Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002652.2 | NP_002643.1 | 3 | 270 | ||
GenBank mRNA | AK312227.1 | - | 3 | 271 | ||
BC010950.1 | - | 3 | 278 | 4250950 | ||
BC010951.1 | - | 3 | 278 | 4295801 | ||
CD723133.1 | - | 3 | 257 | |||
J03460.1 | - | 3 | 259 | |||
Y10179.1 | - | 3 | 270 |