Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002652.2 | NP_002643.1 | 4 | 381 | ||
GenBank mRNA | AK312227.1 | - | 4 | 382 | ||
BC010950.1 | - | 4 | 389 | 4250950 | ||
BC010951.1 | - | 4 | 389 | 4295801 | ||
CD723133.1 | - | 4 | 368 | |||
J03460.1 | - | 4 | 370 | |||
Y10179.1 | - | 4 | 381 |