Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000301.3 | NP_000292.1 | 11 | 1483 | ||
GenBank mRNA | AK298338.1 | - | 7 | 839 | ||
AK308036.1 | - | 10 | 959 | |||
AK312966.1 | - | 11 | 1434 | |||
BC060513.1 | - | 11 | 1409 | 4609795 | ||
K02922.1 | - | 4 | 498 | |||
M74220.1 | - | 11 | 1420 | |||
X05199.1 | - | 11 | 1425 |