Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005030.5 | NP_005021.2 | 3, 4 | 878 | ||
GenBank mRNA | AB209179.1 | - | 2, 3 | 2737 | ||
AK303263.1 | - | 4, 5 | 494 | |||
AK308276.1 | - | 4, 5 | 735 | |||
AK313227.1 | - | 3, 4 | 802 | |||
BC002369.2 | - | 3, 4 | 770 | 2822226 | ||
BC003002.1 | - | 3, 4 | 748 | 2823623 | ||
BC014846.1 | - | 3, 4 | 723 | 3854860 | ||
L19559.1 | - | 3, 4 | 780 | |||
U01038.1 | - | 3, 4 | 780 | |||
X73458.1 | - | 3, 4 | 794 | |||
X75932.1 | - | 3, 4 | 780 |