Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005030.5 | NP_005021.2 | 5 | 1020 | ||
GenBank mRNA | AB209179.1 | - | 4 | 2879 | ||
AK303263.1 | - | 6 | 636 | |||
AK308276.1 | - | 6 | 877 | |||
AK313227.1 | - | 5 | 944 | |||
BC002369.2 | - | 5 | 912 | 2822226 | ||
BC003002.1 | - | 5 | 890 | 2823623 | ||
BC014846.1 | - | 5 | 865 | 3854860 | ||
L19559.1 | - | 5 | 922 | |||
U01038.1 | - | 5 | 922 | |||
X73458.1 | - | 5 | 936 | |||
X75932.1 | - | 5 | 922 |