Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000935.2 | NP_000926.2 | 7, 8 | 1282 | ||
NM_182943.2 | NP_891988.1 | 7, 8 | 1282 | |||
XM_005247535.3 | XP_005247592.1 | 8, 9 | 839 | |||
XM_017006625.1 | XP_016862114.1 | 8, 9 | 793 | |||
GenBank mRNA | AB209879.1 | - | 7, 8 | 954 | ||
AK295084.1 | - | 7, 8 | 873 | |||
BC037169.1 | - | 7, 8 | 953 | 4994235 | ||
U84573.1 | - | 7, 8 | 776 |