Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201564.1 | NP_001188493.1 | 3 | 369 | ||
NM_002669.3 | NP_002660.1 | 3 | 369 | |||
GenBank mRNA | AF044333.1 | - | 2 | 244 | ||
AK002108.1 | - | 3 | 344 | |||
AK304574.1 | - | 3 | 328 | |||
AK307997.1 | - | 3 | 316 | |||
BC020786.1 | - | 3 | 333 | 4809482 | ||
BC106004.1 | - | 3 | 307 | 3889109 | ||
BP314297.1 | - | 3 | 369 |