Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242920.1 | NP_001229849.1 | 9 | 1055 | ||
NM_001242921.1 | NP_001229850.1 | 9 | 967 | |||
NM_006227.3 | NP_006218.1 | 11 | 1340 | |||
NM_182676.2 | NP_872617.1 | 10 | 1184 | |||
GenBank mRNA | AB076694.1 | - | 11 | 1124 | ||
AF052157.1 | - | 8 | 842 | |||
AK092397.1 | - | 11 | 1081 | |||
AK092523.1 | - | 10 | 972 | |||
AK097009.1 | - | 10 | 1638 | |||
AK222690.1 | - | 11 | 1128 | |||
AK289371.1 | - | 10 | 972 | |||
AK293150.1 | - | 9 | 883 | |||
AK299181.1 | - | 9 | 847 | |||
BC005045.1 | - | 10 | 965 | 3510757 | ||
BC019847.1 | - | 11 | 1122 | 4997011 | ||
BC019898.1 | - | 11 | 1115 | 4992839 | ||
L26232.1 | - | 11 | 1130 |