Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163257.1 | NP_001156729.1 | 9 | 2044 | ||
NM_005393.2 | NP_005384.2 | 8 | 1878 | |||
GenBank mRNA | AB033032.1 | - | 8 | 1818 | ||
AF149019.1 | - | 8 | 1818 | |||
AK292535.1 | - | 8 | 1878 | |||
AK295762.1 | - | 8 | 1898 | |||
AK295849.1 | - | 5 | 568 | |||
AK295894.1 | - | 7 | 762 | |||
AK296538.1 | - | 5 | 453 | |||
AK304705.1 | - | 8 | 943 | |||
AK307894.1 | - | 9 | 2021 |