Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007106.3 | NP_009037.1 | 2, 3 | 1275 | ||
GenBank mRNA | AF044221.1 | - | 2, 3 | 234 | ||
AK056580.1 | - | 2, 3 | 1170 | |||
AK307361.1 | - | 2, 3 | 219 | |||
AK309253.1 | - | 2, 3 | 204 | |||
AK311846.1 | - | 2, 3 | 318 | |||
AL080177.1 | - | 2, 3 | 239 | |||
BC044582.1 | - | 2, 3 | 718 | 5262176 | ||
BC059385.1 | - | 2, 3 | 615 | 5287541 | ||
CR533458.1 | - | 2, 3 | 130 |