Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016937.3 | NP_058633.2 | 5, 6 | 464 | ||
XM_005274552.3 | XP_005274609.1 | 5, 6 | 596 | |||
XM_006724499.2 | XP_006724562.1 | 4, 5 | 406 | |||
XM_011545540.2 | XP_011543842.1 | 5, 6 | 551 | |||
XM_011545541.2 | XP_011543843.1 | 5, 6 | 552 | |||
XM_017029594.1 | XP_016885083.1 | 5, 6 | 552 | |||
XM_017029595.1 | XP_016885084.1 | 5, 6 | 552 | |||
XM_017029596.1 | XP_016885085.1 | 5, 6 | 552 | |||
GenBank mRNA | BX648513.1 | - | 5, 6 | 489 | ||
X06745.1 | - | 5, 6 | 465 |