Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291969.1 | NP_001278898.1 | 9 | 2010 | ||
NM_001291970.1 | NP_001278899.1 | 11 | 2298 | |||
NM_006502.2 | NP_006493.1 | 11 | 2369 | |||
XM_005249186.3 | XP_005249243.1 | 10 | 2222 | |||
XM_011514698.2 | XP_011513000.1 | 10 | 2143 | |||
GenBank mRNA | AB024313.1 | - | 11 | 2302 | ||
AF158185.1 | - | 11 | 2230 | |||
AJ420484.1 | - | 8 | 1686 | |||
AK023893.1 | - | 11 | 2360 | |||
AK025631.1 | - | 1697 | ||||
AK122807.1 | - | 8 | 2277 | |||
AK294430.1 | - | 11 | 2350 | |||
AK316151.1 | - | 10 | 2184 | |||
BC015742.1 | - | 11 | 2142 | 4872554 |