Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291970.1 | NP_001278899.1 | 4 | 589 | ||
NM_006502.2 | NP_006493.1 | 4 | 589 | |||
XM_005249186.3 | XP_005249243.1 | 3 | 442 | |||
GenBank mRNA | AB024313.1 | - | 4 | 522 | ||
AF158185.1 | - | 4 | 450 | |||
AK023893.1 | - | 4 | 580 | |||
AK122807.1 | - | 1 | 497 | |||
AK294430.1 | - | 4 | 570 | |||
AK316151.1 | - | 3 | 404 | |||
BC015742.1 | - | 4 | 433 | 4872554 |