Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291969.1 | NP_001278898.1 | 3 | 560 | ||
NM_001291970.1 | NP_001278899.1 | 5 | 919 | |||
NM_006502.2 | NP_006493.1 | 5 | 919 | |||
XM_005249186.3 | XP_005249243.1 | 4 | 772 | |||
XM_011514698.2 | XP_011513000.1 | 4 | 693 | |||
GenBank mRNA | AB024313.1 | - | 5 | 852 | ||
AF158185.1 | - | 5 | 780 | |||
AJ420484.1 | - | 2 | 236 | |||
AK023893.1 | - | 5 | 910 | |||
AK025631.1 | - | 247 | ||||
AK122807.1 | - | 2 | 827 | |||
AK294430.1 | - | 5 | 900 | |||
AK307660.1 | - | 3 | 545 | |||
AK316151.1 | - | 4 | 734 | |||
BC015742.1 | - | 5 | 763 | 4872554 |