Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000938.2 | NP_000929.1 | 13 | 2094 | ||
NM_001303268.1 | NP_001290197.1 | 12 | 1943 | |||
NM_001303269.1 | NP_001290198.1 | 14 | 2272 | |||
GenBank mRNA | AK289823.1 | - | 13 | 1838 | ||
AK294901.1 | - | 12 | 1719 | |||
AK295132.1 | - | 14 | 2043 | |||
AK299090.1 | - | 13 | 1610 | |||
BC023503.2 | - | 13 | 1852 | 3346389 | ||
X63563.1 | - | 13 | 1791 |