Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000940.2 | NP_000931.1 | 6 | 565 | ||
GenBank mRNA | AF320003.1 | - | 6 | 540 | ||
AF329433.1 | - | 6 | 540 | |||
AK096114.1 | - | 1 | 1896 | |||
AK304282.1 | - | 7 | 702 | |||
BC070374.1 | - | 7 | 551 | 4710338 | ||
CD014095.1 | - | 7 | 697 | |||
CD014097.1 | - | 6 | 553 | |||
CD014102.1 | - | 6 | 553 | |||
L48516.1 | - | 6 | 501 |