Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014225.5 | NP_055040.2 | 11 | 1623 | ||
NR_033500.1 | 10 | 1522 | ||||
XM_017026929.1 | XP_016882418.1 | 11 | 1852 | |||
GenBank mRNA | AK090488.1 | - | 11 | 1857 | ||
AK091387.1 | - | 11 | 1227 | |||
AK130000.1 | - | 10 | 1131 | |||
AK290593.1 | - | 11 | 1411 | |||
AK291932.1 | - | 11 | 1406 | |||
AK293174.1 | - | 12 | 1374 | |||
AK293493.1 | - | 10 | 1247 | |||
AK299012.1 | - | 11 | 1278 | |||
AK315700.1 | - | 11 | 1439 | |||
BC001537.2 | - | 11 | 1369 | 2987938 | ||
BC026733.1 | - | 11 | 1386 | |||
CR450340.1 | - | 11 | 1328 | |||
J02902.1 | - | 11 | 1472 |