Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282179.1 | NP_001269108.1 | 8 | 1405 | ||
NM_001282180.1 | NP_001269109.1 | 8 | 1408 | |||
NM_001282181.1 | NP_001269110.1 | 7 | 775 | |||
NM_001282182.1 | NP_001269111.1 | 7 | 730 | |||
NM_006246.3 | NP_006237.1 | 8 | 1408 | |||
NR_104104.1 | 7 | 1211 | ||||
GenBank mRNA | AB209811.1 | - | 8 | 1361 | ||
AB451233.1 | - | 7 | 818 | |||
AB451358.1 | - | 7 | 818 | |||
AK299520.1 | - | 8 | 1345 | |||
AK301796.1 | - | 7 | 775 | |||
AK313500.1 | - | 8 | 1261 | |||
AK316430.1 | - | 7 | 729 | |||
BC013154.1 | - | 7 | 1204 | |||
BC034994.1 | - | 9 | 1402 | |||
BC048305.1 | - | 8 | 1141 | |||
BC064358.1 | - | 8 | 1388 | |||
BC073873.1 | - | 8 | 1360 | |||
BC092477.1 | - | 8 | 1378 | 5267163 | ||
BC093766.1 | - | 8 | 876 | 7939611 | ||
BC101479.1 | - | 8 | 876 | 8068985 | ||
BC143231.1 | - | 8 | 873 | 9051732 | ||
BC143234.1 | - | 8 | 876 | 9051735 | ||
L76703.1 | - | 8 | 1385 | |||
Z69029.1 | - | 8 | 1323 |