Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123355.1 | NP_001116827.1 | 1 | 240 | ||
NM_001123369.1 | NP_001116841.1 | 1 | 240 | |||
NM_002721.4 | NP_002712.1 | 1 | 240 | |||
GenBank mRNA | AF035158.2 | - | 1 | 19 | ||
AK295190.1 | - | 1 | 105 | |||
AK295752.1 | - | 1 | 60 | |||
AK299087.1 | - | 1 | 104 | |||
AK312332.1 | - | 1 | 72 | |||
AK316527.1 | - | 1 | 94 | |||
BC006990.1 | - | 1 | 87 | 3684339 | ||
DB037210.1 | - | 1 | 240 | |||
DC334216.1 | - | 1 | 103 | |||
DC369815.1 | - | 1 | 104 | |||
KU178271.1 | - | 1 | 19 | |||
KU178272.1 | - | 1 | 19 | |||
X92972.2 | - | 1 | 40 |