Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123355.1 | NP_001116827.1 | 8 | 1200 | ||
NM_001123369.1 | NP_001116841.1 | 6 | 1023 | |||
NM_002721.4 | NP_002712.1 | 7 | 1089 | |||
XM_011518847.2 | XP_011517149.1 | 7 | 886 | |||
GenBank mRNA | AF035158.2 | - | 7 | 862 | ||
AK295190.1 | - | 8 | 1065 | |||
AK295752.1 | - | 5 | 701 | |||
AK299087.1 | - | 6 | 887 | |||
AK312332.1 | - | 7 | 921 | |||
AK316527.1 | - | 5 | 735 | |||
BC006990.1 | - | 7 | 936 | 3684339 | ||
BT019708.1 | - | 7 | 868 | |||
KU178271.1 | - | 7 | 868 | |||
KU178272.1 | - | 5 | 660 | |||
X92972.2 | - | 7 | 889 |