Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319214.1 | NP_001306143.1 | 6 | 864 | ||
NM_005040.3 | NP_005031.1 | 7 | 1226 | |||
NM_199418.3 | NP_955450.2 | 8 | 1289 | |||
XM_005274093.1 | XP_005274150.1 | 7 | 956 | |||
GenBank mRNA | AB451270.1 | - | 7 | 1025 | ||
AB451397.1 | - | 7 | 1025 | |||
AK090847.1 | - | 1 | 1522 | |||
AK091786.1 | - | 8 | 1116 | |||
AK302386.1 | - | 6 | 815 | |||
AK312919.1 | - | 7 | 1047 | |||
BC001500.2 | - | 7 | 1044 | 3510081 | ||
L13977.1 | - | 7 | 1054 |