Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKACB SIRNA Assays ›
Gene Symbol:
PRKACB
Gene Name:
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases:
PKA C-beta, PKACB
Chromosome Location:
Chr.1: 84077975 - 84238498 on Build GRCh38
UniGene ID:
Hs.487325
Species:
Human
siRNA ID
HSS108469
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKACB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242857.2
NP_001229786.1
9
739
NM_001242858.2
NP_001229787.1
6
682
NM_001242859.2
NP_001229788.1
8
730
NM_001242860.2
NP_001229789.1
9
874
NM_001242861.2
NP_001229790.1
7
766
NM_001242862.2
NP_001229791.1
5
478
NM_001300915.1
NP_001287844.1
9
874
NM_001300916.1
NP_001287845.1
6
744
NM_001300917.1
NP_001287846.1
6
682
NM_002731.3
NP_002722.1
6
840
NM_182948.3
NP_891993.1
6
744
NM_207578.2
NP_997461.1
6
840
XM_005271015.3
XP_005271072.1
8
691
XM_005271016.1
XP_005271073.1
8
816
XM_005271017.3
XP_005271074.1
7
681
XM_005271018.4
XP_005271075.1
7
1001
XM_005271019.1
XP_005271076.1
6
757
XM_005271020.1
XP_005271077.1
6
724
XM_006710758.1
XP_006710821.1
8
830
XM_017001706.1
XP_016857195.1
9
734
XM_017001707.1
XP_016857196.1
9
734
XM_017001708.1
XP_016857197.1
9
835
XM_017001709.1
XP_016857198.1
8
532
XM_017001710.1
XP_016857199.1
8
830
XM_017001711.1
XP_016857200.1
8
532
XM_017001712.1
XP_016857201.1
8
830
XM_017001713.1
XP_016857202.1
6
782
XM_017001714.1
XP_016857203.1
8
816
XM_017001715.1
XP_016857204.1
8
816
XM_017001716.1
XP_016857205.1
7
684
XM_017001717.1
XP_016857206.1
7
1005
XM_017001718.1
XP_016857207.1
6
659
XM_017001719.1
XP_016857208.1
6
757
XM_017001720.1
XP_016857209.1
6
757
GenBank mRNA
AB209189.1
-
9
810
AB451239.1
-
6
489
AB451364.1
-
6
489
AK091420.1
-
7
440
AK296482.1
-
9
541
AK304322.1
-
6
669
AK304375.1
-
6
484
AK314550.1
-
6
736
AK316115.1
-
5
451
BC016285.1
-
6
671
3916100
BI598037.1
-
6
555
BM905428.1
-
6
626
BX537705.1
-
6
721
BX641026.1
-
6
720
BX647515.1
-
9
819
CR936631.1
-
8
533
DA228647.1
-
5
479
M34181.1
-
6
536
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
stimulatory C-type lectin receptor signaling pathway
renal water homeostasis
protein phosphorylation
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
blood coagulation
activation of protein kinase A activity
lipoprotein metabolic process
negative regulation of meiotic cell cycle
regulation of protein processing
cellular response to glucagon stimulus
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
protein binding
ATP binding
kinase activity
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0