Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276289.1 | NP_001263218.1 | 8 | 992 | ||
NM_001276290.1 | NP_001263219.1 | 6 | 689 | |||
NM_001278433.1 | NP_001265362.1 | 7 | 862 | |||
NM_002734.4 | NP_002725.1 | 7 | 859 | |||
NM_212471.2 | NP_997636.1 | 7 | 1052 | |||
NM_212472.2 | NP_997637.1 | 7 | 890 | |||
XM_011524983.2 | XP_011523285.1 | 7 | 955 | |||
XM_011524984.2 | XP_011523286.1 | 8 | 893 | |||
XM_011524985.2 | XP_011523287.1 | 8 | 1187 | |||
GenBank mRNA | A12295.1 | - | 7 | 785 | ||
AK097580.1 | - | 6 | 1262 | |||
AK124586.1 | - | 7 | 778 | |||
AK312307.1 | - | 7 | 788 | |||
AL535889.3 | - | 7 | 830 | |||
AW732104.1 | - | 5 | 526 | |||
BC036285.1 | - | 7 | 811 | 4340015 | ||
BC093042.1 | - | 7 | 800 | 6514677 | ||
CR749311.1 | - | 8 | 921 | |||
M18468.1 | - | 7 | 786 | |||
M33336.1 | - | 7 | 770 | |||
S54707.1 | - | 3 | 215 |