Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164758.1 | NP_001158230.1 | 4, 5 | 552 | ||
NM_001164759.1 | NP_001158231.1 | 4, 5 | 487 | |||
NM_001164760.1 | NP_001158232.1 | 4, 5 | 597 | |||
NM_001164761.1 | NP_001158233.1 | 4, 5 | 618 | |||
NM_001164762.1 | NP_001158234.1 | 4, 5 | 558 | |||
NM_002735.2 | NP_002726.1 | 4, 5 | 576 | |||
XM_011515446.2 | XP_011513748.1 | 3, 4 | 534 | |||
GenBank mRNA | AK098822.1 | - | 3, 4 | 518 | ||
AK289464.1 | - | 4, 5 | 499 | |||
AK297337.1 | - | 4, 5 | 528 | |||
AK315951.1 | - | 4, 5 | 588 | |||
AK315990.1 | - | 4, 5 | 548 | |||
AL833563.1 | - | 4, 5 | 518 | |||
BC026734.1 | - | 4, 5 | 526 | 5141110 | ||
BC036828.1 | - | 4, 5 | 549 | 5247772 | ||
DA210679.1 | - | 4, 5 | 488 | |||
M65066.1 | - | 3, 4 | 441 |