Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002738.6 | NP_002729.2 | 9 | 1208 | ||
NM_212535.2 | NP_997700.1 | 9 | 1208 | |||
GenBank mRNA | BC036472.1 | - | 9 | 1168 | 5260547 | |
M13975.1 | - | 9 | 1075 | |||
X06318.1 | - | 9 | 1147 | |||
X07109.1 | - | 9 | 1147 |