Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006255.4 | NP_006246.2 | 3 | 1099 | ||
XM_011536954.2 | XP_011535256.1 | 3 | 282 | |||
XM_011536955.1 | XP_011535257.1 | 3 | 215 | |||
XM_011536956.1 | XP_011535258.1 | 3 | 981 | |||
XM_011536957.1 | XP_011535259.1 | 3 | 981 | |||
XM_017021458.1 | XP_016876947.1 | 3 | 433 | |||
XM_017021459.1 | XP_016876948.1 | 3 | 981 | |||
GenBank mRNA | AB451313.1 | - | 3 | 440 | ||
AK290183.1 | - | 3 | 818 | |||
AK296158.1 | - | 2 | 113 | |||
AX775931.1 | - | 3 | 175 | |||
AX775933.1 | - | 3 | 828 | |||
BC037268.1 | - | 3 | 828 | 4822159 | ||
M55284.1 | - | 3 | 603 |