Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002741.3 | NP_002732.3 | 10 | 1606 | ||
NM_213560.1 | NP_998725.1 | 10 | 1473 | |||
XM_011528128.1 | XP_011526430.1 | 10 | 1489 | |||
GenBank mRNA | AK123007.1 | - | 10 | 1487 | ||
AK292130.1 | - | 10 | 1470 | |||
AK313886.1 | - | 10 | 1446 | |||
BC018575.1 | - | 4 | 324 | |||
BC040061.1 | - | 10 | 1603 | 5752583 | ||
BC094766.1 | - | 10 | 1637 | 30342125 | ||
D26181.1 | - | 10 | 1474 | |||
S75546.1 | - | 10 | 1522 | |||
U33053.1 | - | 10 | 1522 |