Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282480.1 | NP_001269409.1 | 7 | 588 | ||
NM_001282481.1 | NP_001269410.1 | 6 | 462 | |||
NM_001282483.1 | NP_001269412.1 | 6 | 462 | |||
NM_006259.2 | NP_006250.1 | 11 | 1513 | |||
XM_005263126.3 | XP_005263183.1 | 11 | 1948 | |||
XM_017008413.1 | XP_016863902.1 | 11 | 1950 | |||
XM_017008414.1 | XP_016863903.1 | 11 | 1945 | |||
XM_017008415.1 | XP_016863904.1 | 11 | 1355 | |||
XM_017008416.1 | XP_016863905.1 | 11 | 1867 | |||
XM_017008418.1 | XP_016863907.1 | 10 | 1331 | |||
GenBank mRNA | AK296979.1 | - | 6 | 449 | ||
AK316140.1 | - | 7 | 588 | |||
D70899.1 | - | 10 | 1318 | |||
DA462272.1 | - | 6 | 462 | |||
X94612.1 | - | 10 | 1332 |