Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135044.1 | NP_001128516.1 | 5 | 701 | ||
NM_001308244.1 | NP_001295173.1 | 4 | 477 | |||
NM_002752.4 | NP_002743.3 | 5 | 701 | |||
NM_139068.2 | NP_620707.1 | 5 | 701 | |||
NM_139069.2 | NP_620708.1 | 5 | 701 | |||
NM_139070.2 | NP_620709.1 | 5 | 701 | |||
XM_005265940.4 | XP_005265997.1 | 6 | 834 | |||
XM_006714891.2 | XP_006714954.1 | 6 | 759 | |||
XM_017009638.1 | XP_016865127.1 | 6 | 670 | |||
XM_017009639.1 | XP_016865128.1 | 7 | 927 | |||
XM_017009640.1 | XP_016865129.1 | 6 | 818 | |||
XM_017009641.1 | XP_016865130.1 | 6 | 768 | |||
XM_017009642.1 | XP_016865131.1 | 5 | 597 | |||
XM_017009643.1 | XP_016865132.1 | 5 | 597 | |||
GenBank mRNA | AB451302.1 | - | 4 | 430 | ||
AB451355.1 | - | 4 | 430 | |||
AK289638.1 | - | 5 | 597 | |||
BC032539.1 | - | 4 | 491 | 5528624 | ||
CR536580.1 | - | 4 | 430 | |||
EU927388.1 | - | 4 | 450 | |||
HM036706.1 | - | 4 | 450 | |||
HM036707.1 | - | 4 | 450 | |||
HM036708.1 | - | 4 | 450 | |||
HM036709.1 | - | 5 | 691 | |||
L31951.1 | - | 4 | 488 | |||
U09759.1 | - | 4 | 581 | |||
U34821.1 | - | 4 | 474 | |||
U35002.1 | - | 4 | 452 | |||
U35003.1 | - | 4 | 464 |