Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000313.3 | NP_000304.2 | 11 | 1549 | ||
NM_001314077.1 | NP_001301006.1 | 12 | 1636 | |||
GenBank mRNA | AK292994.1 | - | 11 | 1355 | ||
AK303895.1 | - | 4 | 476 | |||
BC015801.1 | - | 11 | 1525 | 3909023 | ||
KU178283.1 | - | 11 | 1208 | |||
KU178284.1 | - | 12 | 1304 | |||
M14338.1 | - | 11 | 1376 | |||
M15036.1 | - | 11 | 1354 | |||
X12892.1 | - | 11 | 1376 | |||
Y00692.1 | - | 11 | 1331 |