Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRPSAP2 SIRNA Assays ›
Gene Symbol:
PRPSAP2
Gene Name:
phosphoribosyl pyrophosphate synthetase associated protein 2
Gene Aliases:
PAP41
Chromosome Location:
Chr.17: 18856299 - 18931287 on Build GRCh38
UniGene ID:
Hs.632236
Species:
Human
siRNA ID
HSS108608
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRPSAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243936.1
NP_001230865.1
3
313
NM_001243940.1
NP_001230869.1
2
182
NM_001243941.1
NP_001230870.1
2
280
NM_001243942.1
NP_001230871.1
3
313
NM_002767.3
NP_002758.1
3
376
XM_005256725.2
XP_005256782.1
2
230
XM_005256726.1
XP_005256783.1
2
264
XM_005256729.4
XP_005256786.1
2
179
XM_017024864.1
XP_016880353.1
3
240
XM_017024865.1
XP_016880354.1
4
449
XM_017024866.1
XP_016880355.1
4
383
XM_017024867.1
XP_016880356.1
2
217
XM_017024868.1
XP_016880357.1
3
1069
XM_017024869.1
XP_016880358.1
3
348
XM_017024870.1
XP_016880359.1
3
263
XM_017024871.1
XP_016880360.1
3
326
XM_017024872.1
XP_016880361.1
4
462
XM_017024873.1
XP_016880362.1
3
311
XM_017024874.1
XP_016880363.1
3
383
XM_017024875.1
XP_016880364.1
3
256
XM_017024876.1
XP_016880365.1
2
234
GenBank mRNA
AB007851.1
-
2
141
AK303911.1
-
3
259
AK304544.1
-
2
165
AK313087.1
-
3
313
AL535304.3
-
3
273
BC101670.1
-
2
102
8069176
BC101672.1
-
2
102
8069178
BC106050.1
-
3
292
6176727
BC143475.1
-
2
102
9051983
BX648850.1
-
3
262
CR457082.1
-
1
54
DA257655.1
-
2
264
DC424578.1
-
3
376
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
nucleoside metabolic process
nucleotide biosynthetic process
negative regulation of catalytic activity
bone development
magnesium ion binding
ribose phosphate diphosphokinase activity
enzyme inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-4zcjp:80/100.66.78.62:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline