Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005045.3 | NP_005036.2 | 65 | 10500 | ||
NM_173054.2 | NP_774959.1 | 64 | 10494 | |||
GenBank mRNA | AK093843.1 | - | 1 | 1568 | ||
AK293863.1 | - | 22 | 3436 | |||
AK297459.1 | - | 15 | 2064 | |||
U79716.1 | - | 65 | 10515 |