Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005045.3 | NP_005036.2 | 35 | 5428 | ||
NM_173054.2 | NP_774959.1 | 35 | 5428 | |||
GenBank mRNA | U79716.1 | - | 35 | 5443 |