Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012964.2 | NP_001012982.1 | 4 | 756 | ||
NM_001012965.2 | NP_001012983.1 | 3 | 708 | |||
NM_001319948.1 | NP_001306877.1 | 4 | 402 | |||
NM_001319949.1 | NP_001306878.1 | 3 | 599 | |||
NM_002774.3 | NP_002765.1 | 5 | 559 | |||
GenBank mRNA | AF013988.1 | - | 5 | 461 | ||
AK314897.1 | - | 5 | 464 | |||
AY279383.1 | - | 4 | 403 | |||
AY318867.1 | - | 4 | 565 | |||
AY318868.1 | - | 3 | 394 | |||
AY318869.1 | - | 4 | 551 | |||
AY318870.1 | - | 3 | 503 | |||
BC015525.2 | - | 4 | 532 | 3852344 | ||
BT006852.1 | - | 3 | 315 | |||
D78203.1 | - | 5 | 470 | |||
DQ223012.1 | - | 3 | 407 | |||
U62801.1 | - | 5 | 560 |