Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012964.2 | NP_001012982.1 | 4, 5 | 887 | ||
NM_001012965.2 | NP_001012983.1 | 3, 4 | 839 | |||
NM_001319948.1 | NP_001306877.1 | 4, 5 | 533 | |||
NM_001319949.1 | NP_001306878.1 | 3, 4 | 730 | |||
NM_002774.3 | NP_002765.1 | 5, 6 | 690 | |||
GenBank mRNA | AF013988.1 | - | 5, 6 | 592 | ||
AK314897.1 | - | 5, 6 | 595 | |||
AY279383.1 | - | 4, 5 | 534 | |||
AY318867.1 | - | 4, 5 | 696 | |||
AY318868.1 | - | 3, 4 | 525 | |||
AY318869.1 | - | 4, 5 | 682 | |||
AY318870.1 | - | 3, 4 | 634 | |||
BC015525.2 | - | 4, 5 | 663 | 3852344 | ||
BT006852.1 | - | 3, 4 | 446 | |||
D78203.1 | - | 5, 6 | 601 | |||
DQ223012.1 | - | 3, 4 | 538 | |||
U62801.1 | - | 5, 6 | 691 |