Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000021.3 | NP_000012.1 | 12 | 1637 | ||
NM_007318.2 | NP_015557.2 | 12 | 1625 | |||
XM_005267864.2 | XP_005267921.1 | 12 | 1631 | |||
XM_005267866.1 | XP_005267923.1 | 12 | 1620 | |||
XM_011536971.2 | XP_011535273.1 | 12 | 1529 | |||
XM_011536972.2 | XP_011535274.1 | 12 | 1893 | |||
XM_011536973.1 | XP_011535275.1 | 12 | 1881 | |||
XM_011536974.1 | XP_011535276.1 | 12 | 1522 | |||
GenBank mRNA | AK122722.1 | - | 10 | 2906 | ||
AK223173.1 | - | 10 | 1367 | |||
AK303876.1 | - | 8 | 1012 | |||
AK312531.1 | - | 12 | 1621 | |||
BC011729.2 | - | 12 | 1606 | 4423098 | ||
KU178285.1 | - | 10 | 1341 | |||
KU178286.1 | - | 10 | 1353 | |||
KU178287.1 | - | 5 | 572 | |||
L42110.1 | - | 12 | 1601 | |||
L76517.1 | - | 12 | 1894 | |||
U40379.1 | - | 10 | 1341 | |||
U40380.1 | - | 11 | 1433 |