Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270481.1 | NP_001257410.1 | 1 | 131 | ||
NM_002798.2 | NP_002789.1 | 1 | 131 | |||
GenBank mRNA | AK311768.1 | - | 1 | 94 | ||
BC000835.2 | - | 1 | 98 | 3456009 | ||
BF671774.1 | - | 1 | 132 | |||
BM553399.1 | - | 1 | 78 | |||
BQ691763.1 | - | 1 | 113 | |||
BT009809.1 | - | 1 | 80 | |||
BU848746.1 | - | 1 | 63 | |||
CR457065.1 | - | 1 | 80 | |||
D29012.1 | - | 1 | 89 |