Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002803.3 | NP_002794.1 | 9 | 956 | ||
GenBank mRNA | AK298821.1 | - | 8 | 717 | ||
AK312648.1 | - | 9 | 807 | |||
BC002589.2 | - | 9 | 800 | 3161790 | ||
D11094.1 | - | 9 | 834 |