Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199163.1 | NP_001186092.1 | 4, 5 | 484 | ||
NM_002805.5 | NP_002796.4 | 4, 5 | 349 | |||
GenBank mRNA | AF035309.1 | - | 427 | |||
AK095836.1 | - | 3, 4 | 1583 | |||
AK290758.1 | - | 4, 5 | 447 | |||
AK291878.1 | - | 4, 5 | 321 | |||
BC001932.1 | - | 4, 5 | 300 | 2820259 | ||
BC002367.2 | - | 4, 5 | 279 | 2821416 | ||
BC127888.1 | - | 2, 3 | 191 | |||
BF245599.1 | - | 333 | ||||
D44467.1 | - | 4, 5 | 279 | |||
L38810.1 | - | 4, 5 | 286 |