Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002806.3 | NP_002797.3 | 4, 5 | 294 | ||
GenBank mRNA | AF006305.1 | - | 4, 5 | 267 | ||
AK299155.1 | - | 3, 4 | 208 | |||
AK313670.1 | - | 4, 5 | 262 | |||
BC005390.1 | - | 4, 5 | 272 | 3996906 | ||
BM788029.1 | - | 4, 5 | 294 | |||
BT006843.1 | - | 4, 5 | 252 | |||
CR456709.1 | - | 4, 5 | 252 | |||
D78275.1 | - | 4, 5 | 268 | |||
FJ224299.1 | - | 4, 5 | 268 |