Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191037.1 | NP_001177966.1 | 4 | 426 | ||
NM_002807.3 | NP_002798.2 | 4 | 426 | |||
XM_017004517.1 | XP_016860006.1 | 4 | 426 | |||
GenBank mRNA | AK312528.1 | - | 4 | 383 | ||
BC001053.2 | - | 4 | 383 | |||
BC014013.2 | - | 4 | 395 | |||
BC020223.1 | - | 4 | 373 | |||
BC032445.1 | - | 4 | 398 | |||
BC039845.1 | - | 4 | 349 | |||
BC047897.1 | - | 4 | 348 | |||
BC064398.1 | - | 4 | 397 | |||
BC073833.1 | - | 4 | 350 | |||
BC094720.1 | - | 4 | 412 | 4826340 | ||
BC112344.1 | - | 4 | 387 | 40008454 | ||
BC114434.1 | - | 4 | 387 | 40008444 | ||
D44466.1 | - | 4 | 344 | |||
DA385817.1 | - | 4 | 426 |