Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261400.2 | NP_001248329.1 | 3, 4 | 463 | ||
NM_002813.6 | NP_002804.2 | 5, 6 | 778 | |||
NR_048555.2 | 4, 5 | 566 | ||||
GenBank mRNA | AK055339.1 | - | 1, 2 | 1734 | ||
AK315389.1 | - | 5, 6 | 747 | |||
BC002383.2 | - | 5, 6 | 738 | 2961610 | ||
BC004184.2 | - | 5, 6 | 738 | 2961610 | ||
BC004213.2 | - | 5, 6 | 738 | 2961610 | ||
BM850893.1 | - | 4, 5 | 487 | |||
CB111716.1 | - | 3, 4 | 408 |